Wavelength calibration solution for ICP-OES and MP-AES
Lieferant: VARIAN
6610030100
VARI6610030100EA 651 EUR
VARI6610030100
Wavelength calibration solution for ICP-OES and MP-AES
Standards Kationischer Standards Mehrelement-Standards
500 ml solution contains 5 mg/l Al, As, Ba,Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn and 50 mg/l K in 5 % HNO3 + tr HF.
  • Ready to use
  • Packaged in pre-leached HDPE bottles, secured with a tamper-evident seal
Lieferumfang: Geliefert mit Analysenzertifikat.