SI plates, Bond Elut™
Lieferant: VARIAN
7542502C 7541301C
VARI7542502CEA 506 EUR
VARI7542502C VARI7541301C
SI plates, Bond Elut™
Mikrotiterplatten Festphasenextraktionsplatten
Native silica is generally regarded as the most polar SPE sorbent available. Bond Elut SI is particularly effective at separating compounds with a very similar structure.
  • Highly polar phase retains polar molecules from nonpolar matrices
  • High purity silica
  • Separate compounds with very similar structures
Native silica is generally regarded as the most polar SPE sorbent available. Bond Elut SI is particularly effective at separating compounds with a very similar structure. Applying the analytes in a non-polar solvent, then increasing the solvent polarity by increasing the concentration of a polar modifier, such as THF or ethyl acetate, delivers effective separations.