Robotic OMIX™ tips for the Hamilton Microlab® Star Line
Lieferant: VARIAN
A57403
VARIA57403EA 0 EUR
VARIA57403
Robotic OMIX™ tips for the Hamilton Microlab® Star Line
Pipettenspitzen
These tips offer excellent versatility and end-user productivity enhancements. Processing of 96 samples can be reduced to just a few minutes in certain applications. The tips have an operating volume of 300 µl, allowing flexibility in sample size.