Vacuum trap assembly, A584
Lieferant: VARIAN
A584
VARIA584EA 74.1 EUR
VARIA584
Vacuum trap assembly, A584
Vakuumfallen
This vacuum trap eliminates the possibility of drawing fluids into the vacuum pump that may be detrimental to it.