Mobile phase inlet filter, Hastelloy® C
Lieferant: VARIAN
AL9402
VARIAL9402EA 0 EUR
VARIAL9402
Mobile phase inlet filter, Hastelloy® C
Filter Flüssigkeits-Prozessfilter
This mobile phase inlet filter operates up to 500 °C and is compatible with halogenated solvents.