Fused silica cutting tools
Lieferant: VARIAN
CP22672 CP22674 CP22675
VARICP22672EA 0 EUR
VARICP22672 VARICP22674 VARICP22675
Fused silica cutting tools
Schneidewerkzeuge
This high quality, long life tungsten carbide glass knife produces perfect cuts, and is especially suitable for use with CP-Quick-Seal connectors.