Pre-prepared polystyrene kits, EasiCal™
Lieferant: VARIAN
PL2010-0505
PL2010-0501
PL2010-0601
PL2010-0605
VARIPL2010-0505EA
1590
EUR
VARIPL2010-0505
VARIPL2010-0501
VARIPL2010-0601
VARIPL2010-0605
Pre-prepared polystyrene kits, EasiCal™
Standards
Gaschromatographie-Standards
The EasiCal system for organic solvents consists of two different combs, each with ten detachable spatulas supporting a mixture of five polymer standards with MWs selected to provide equidistant calibration points for greater accuracy.
- Easy three step process with no fuss
- Cost effective
- Only two injections for improved productivity
- Pre-prepared for easy use
The thin film of polymer (approximately 5 mg) on the tip of the PTFE spatulas rapidly dissolves when immersed in eluent to provide two GPC/SEC calibration solutions.