Pre-prepared polystyrene kits, EasiCal™
Lieferant: VARIAN
PL2010-0505 PL2010-0501 PL2010-0601 PL2010-0605
VARIPL2010-0505EA 1590 EUR
VARIPL2010-0505 VARIPL2010-0501 VARIPL2010-0601 VARIPL2010-0605
Pre-prepared polystyrene kits, EasiCal™
Standards Gaschromatographie-Standards
The EasiCal system for organic solvents consists of two different combs, each with ten detachable spatulas supporting a mixture of five polymer standards with MWs selected to provide equidistant calibration points for greater accuracy.
  • Easy three step process with no fuss
  • Cost effective
  • Only two injections for improved productivity
  • Pre-prepared for easy use
The thin film of polymer (approximately 5 mg) on the tip of the PTFE spatulas rapidly dissolves when immersed in eluent to provide two GPC/SEC calibration solutions.