Seal kit for liquid head pump
Lieferant: VARIAN
R007105625
VARIR007105625EA 0 EUR
VARIR007105625
Seal kit for liquid head pump
Chromatographie-Zubehör Chromatographie-Ersatzteile
Seal kits are available in various capacity based on the pump head sizes.
Lieferumfang: Seal kit includes high pressure seals, washing section seals and O-rings.