Analytical HPLC columns, Microsorb™ Si
Lieferant: VARIAN
R0089100C5 R0089100D5 R0089100E3
VARIR0089100C5EA 846 EUR
VARIR0089100C5 VARIR0089100D5 VARIR0089100E3
Analytical HPLC columns, Microsorb™ Si
Chromatographiesäulen
Microsorb™ Silica is ideal for the separation of low molecular weight compounds that dissolve in organic solvents. Separations on silica columns depend upon the difference in orientation, type and number of functional groups associated with the compounds in the sample.