Immunohistochemistry reagents, Horseradish Peroxidase Avidin D
Lieferant: Vector Laboratories
A-2004-5
VECTA-2004-5EA 436 EUR
VECTA-2004-5
Immunohistochemistry reagents, Horseradish Peroxidase Avidin D
Proteinkonjugate und Markierungen
Horseradish peroxidase avidin D is produced by our own coupling procedure which preserves the high specific activity of the peroxidase. Using biotinylated primary or secondary intermediates and peroxidase-labelled avidin D, antigens can be localised in histological sections, cytospin preparations, or smears. In addition, sensitive detection systems for a variety of solid-phase assays utilising microtitration plates or nitrocellulose have been developed using this conjugate.
Horseradish peroxidase avidin D is offered as a concentrate (5 mg/ml).