Immunoperoxidase systems, VECTASTAIN® Elite® ABC Kit (Standard)
Lieferant: Vector Laboratories
PK-6100
VECTPK-6100EA 979 EUR
VECTPK-6100
Immunoperoxidase systems, VECTASTAIN® Elite® ABC Kit (Standard)
Proteinkonjugate und Markierungen
The VECTASTAIN® Elite® ABC kit is supplied without biotinylated secondary antibodies or blocking serum. The 'Standard' kit consists of Elite® A and B reagents, which can be used with any biotinylated primary or secondary antibody, biotinylated nucleic acid probe or other biotinylated molecule.
  • Suitable for use on frozen or paraffin-embedded tissues, cell smears, or cytospin preparations
  • Offers low background and amplified signal compared to conventional covalent conjugates
  • Stock solutions packaged in convenient dropper bottles and each kit includes mixing bottles to prepare the working solutions
  • Detailed instructions for use included
Bestellinformation: Each kit contains sufficient reagents to prepare 110 ml of each working solution (500 to 1000 sections).