L(-)-Fucose

Lieferant: Vector Laboratories

Synonyme: (-)-Fucose, 6-Deoxy-L-Galactose, (2S,3R,4R,5S)-2,3,4,5-Tetrahydroxyhexanal

S-9007
VECTS-9007EA 0 EUR
VECTS-9007
L(-)-Fucose
L(-)-Fucose
Formel: C₆H₁₂O₅
Molecular Weight: 164,16 g/mol
MDL: MFCD00069812
CAS-Nummer: 2438-80-4
EINECS: 219-452-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR