3-Amino-9-ethylcarbazol (AEC)

Lieferant: Vector Laboratories
Gefahr

Synonyme: 9-Ethylcarbazol-3-ylamin, AEC

SK-4205
VECTSK-4205EA 451 EUR
VECTSK-4205
3-Amino-9-ethylcarbazol (AEC)
3-Amino-9-ethyl carbazole (AEC)
Formel: C₁₄H₁₄N₂
Molecular Weight: 210.28 g/mol
Schmelzpunkt: 113…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004964
CAS-Nummer: 132-32-1
EINECS: 205-057-7
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR