Biopharmaceutical tubing C-Flex®, Formulation 072
Lieferant: Saint Gobain Life Sciences
072-250-2
VERN072-250-2EA 171 EUR
VERN072-250-2
Biopharmaceutical tubing C-Flex®, Formulation 072
Schläuche Transferschlauch
Thermoplastic elastomer, opaque.
  • Manufactured under strict GMPs guidelines
  • Non-pyrogenic, non-cytotoxic, non-hemolytic
  • Chemically resistant to concentrated acids and alkalis
  • Significantly less permeable than silicone
  • Low platelet adhesion
  • Low protein binding
  • Ultra-smooth inner bore
  • Lot traceable
C-Flex® is a unique, patented thermoplastic elastomer specifically designed to meet the critical demands of the pharmaceutical, research, biopharmaceutical and diagnostics industries. C-Flex® biopharmaceutical tubing has been used by many of the world’s leading biotech and pharmaceutical processing companies. Each coil of C-Flex® tubing is extruded to precise Int.Ø, Ext.Ø and wall dimensions. All tubing is formulated to meet the standards of the biopharmaceutical industry and is QA tested before leaving the Saint-Gobain Performance Plastics production facility.
Temperature resistance: −45 to +135 °C
Hardness (Shore durometer A): 60
Standards: Complies with USP Class VI standards.