KryoSure® Cryopreservation bags safely freeze and protect biologics at temperatures down to, and even beyond, liquid nitrogen temperatures (–196 °C). All KryoSure® bags are made from the highest quality USP Class VI Fluorinated Ethylene Propylene (FEP) material.
- Full extractables report per BioPhorum Operations Group (BPOG) protocol
- Extreme operating temperature range
- Technical dossier available
- Chemical and biologically inert
- High purity
- Transparent
- Closed system
Applications: Cryopreservation bags designed as a closed system for freezing, thawing, and transfer of sterile contents.
Bestellinformation: Normally not stocked, built to order sold as 10 individually packaged sterile bags per pack.