KryoSure® Cryopreservation Bag
Lieferant: Saint Gobain Life Sciences
20-F
VERN20-FEA 8810 EUR
VERN20-F
KryoSure® Cryopreservation Bag
Beutel Medium-Beutel
KryoSure® Cryopreservation bags safely freeze and protect biologics at temperatures down to, and even beyond, liquid nitrogen temperatures (–196 °C). All KryoSure® bags are made from the highest quality USP Class VI Fluorinated Ethylene Propylene (FEP) material.
  • Full extractables report per BioPhorum Operations Group (BPOG) protocol
  • Extreme operating temperature range
  • Technical dossier available
  • Chemical and biologically inert
  • High purity
  • Transparent
  • Closed system
Applications: Cryopreservation bags designed as a closed system for freezing, thawing, and transfer of sterile contents.
Bestellinformation: Normally not stocked, built to order sold as 10 individually packaged sterile bags per pack.