Versilic™ Stoppers
Lieferant: Saint Gobain Life Sciences
263034
VERN263034EA 195 EUR
VERN263034
Versilic™ Stoppers
Stopfen Silikonstopfen
Use these stoppers for biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus. Saint Gobain Life Sciences Versilic™ Silicone Stoppers are particularly suited for all types of tissue culture work.
  • Biologically neutral and non-toxic silicone caps for closing tubes and flasks
  • Odourless, tasteless, non-adherent and resistant to aging
  • Non toxic, biologically neutral
  • FDA/BGA approved
  • For test tubes
  • Easy to install
Hardness: 45 (shore A)