Tygon S3™ B-44-5X IB Tubings
Lieferant: Saint Gobain Life Sciences
AMH00018 AMH00029 AMH00038 AMH00046 AMH00053 AMH00064 AMH00070 AMH00074
VERNAMH00018EA 365 EUR
VERNAMH00018 VERNAMH00029 VERNAMH00038 VERNAMH00046 VERNAMH00053 VERNAMH00064 VERNAMH00070 VERNAMH00074
Tygon S3™ B-44-5X IB Tubings
Schläuche Transferschlauch
Tygon S3™ B-44-5X IB tubing from Saint-Gobain is clear and flexible with a non-porous bore. It offers outstanding resistance to harsh alkaline cleaners and is unaffected by commonly used sanitisers. Designed specifically for food and beverage dispensing applications, its added reinforcement withstands up to four times the pressure of non-reinforced tubing.
  • All layers compliant with FDA food contact regulations
  • Handles four times the pressure of non-reinforced tubing
  • Improved flexibility at sub-zero temperatures
  • Smooth, non-porous bore resists particle entrapment
  • Clear wall enables visualisation of fluid flow
  • Connects easily to ReSeal® sanitary fittings
Lightweight and easy to handle, Tygon S3 B-44-5X IB tubing goes into service quickly. It readily bends around corners and obstructions, requiring minimal couplings and fittings. Its flexibility can save up to 30% the footage and much of the labour required to install rigid stainless steel, glass tubing or piping. It is ideal for applications such as water lines for beverage dispensing equipment, ice cream dispensing equipment, frozen beverages dispensing machines (smoothies, shakes, slushies).
Hardness: 64 (shore A)
Standards: NSF-51; NSF-61, 3-A Sanitary Standard 20-27; FDA food additive regulations (21CFR 170-199).