Schläuche, Tygon®, A-60-F IB
Lieferant: Saint Gobain Life Sciences
APW00038 APW00019 APW00029 APW00046 APW00054 APW00064
VERNAPW00038EA 0 EUR
VERNAPW00038 VERNAPW00019 VERNAPW00029 VERNAPW00046 VERNAPW00054 VERNAPW00064
Schläuche, Tygon®, A-60-F IB
Schläuche Transferschlauch
Tygon® A-60-F IB is a reinforcement embedded tubing within its walls to withstand elevated pressure and to withstand extreme temperatures from –24 to 135 °C. It will not crack or deteriorate when used in demanding food and beverage applications.
  • Long flex life in peristaltic pumps
  • Compatible with virtually all common sanitizers and cleaners
  • Ozone and UV light resistant
  • Repeatedly autoclavable
  • Chemically compatible with a wide range of food types and cleaners
Tubing provides resistance to many harsh food media, including some animal and vegetable oils. Ideal for use in clean-in-place and steam-in-place cleaning and sterilisation systems.
Temperature resistance: –60 to +135 °C
Hardness (Shore durometer A): 61
Density: 0,98
Standards: Meets FDA 21 CFR, 177.2600, NSF standards 51 and 3-A sanitary standards.