Schläuche, Tygon® 3370 I.B.
Lieferant: Saint Gobain Life Sciences
T1316-63 T1312-43 T1308-43 T1306-43 T1320-63 T1324-63 T1332-63 T1340-66
VERNT1316-63EA 2890 EUR
VERNT1316-63 VERNT1312-43 VERNT1308-43 VERNT1306-43 VERNT1320-63 VERNT1324-63 VERNT1332-63 VERNT1340-66
Schläuche, Tygon® 3370 I.B.
Schläuche Transferschlauch
Braid reinforced for increased pressure resistance, TYGON® 3370 I.B. silicone hose is frequently specified in the most demanding applications requiring sanitary transfer of fluids.
  • Consistently smooth inner surface limits particle entrapment
  • Platinum cured to minimize extractables
  • Tough braid reinforcement permits use under elevated working pressures
  • Withstands repeated CIP and SIP cleaning and sterilization
Its smooth inner surface reduces the risk of particle entrapment and inhibits excessive residue and microscopic bacterial build-up; cleaning and sterilization cycles may become more effective as a result. Additionally, an improvement in fluid flow characteristics may occur from the reduced surface area and lowered absorption of fluids to the wall.
TYGON® 3370 I.B. can easily withstand repeated SIP and CIP cleaning and sterilization cycles, making it ideal for repeat-use applications. Its flexibility, durability, and chemical and temperature resistance provide a unique combination of characteristics required in many pharmaceutical, biotechnology, laboratory, food, beverage, cosmetic, environmental, and industrial applications.
Temperature resistance: −80 to +160 °C
Hardness (Shore durometer A): 70
Density: 1,18