Blaulicht LED-PAD
Lieferant: Vilber Lourmat
213120171
VILB213120171EA 1840 EUR
VILB213120171
Blaulicht LED-PAD
Transilluminatoren Blaulicht-Transilluminatoren
The Skylight Super-Blue transilluminator eliminates the damage caused by UV light on DNA and RNA gels. It also improves cloning efficiency dramatically by eliminating the effects of UV induced nicking or crosslinking, often encountered during the purification of DNA from gels for further use.