UV filtered lamps for fluorescence
Lieferant: Vilber Lourmat
311100631 311100611 311100651 311101511 311101531 311121511 311121521 311121531 311121551 311121561 311121571
VILB311100631EA 548 EUR
VILB311100631 VILB311100611 VILB311100651 VILB311101511 VILB311101531 VILB311121511 VILB311121521 VILB311121531 VILB311121551 VILB311121561 VILB311121571
UV filtered lamps for fluorescence
Beleuchtung UV-Lampen
Lamps are available in single or combined versions.
  • Exclusive UV Master® technology for high UV output
  • Ondulex® reflector for optimum radiation
  • Easy to handle thanks to a table support, handle and wall hook (optional)
The filter minimises white light interference allowing improved detection of weak fluorescence.