Négatoscopes, CN-6
Lieferant: Vilber Lourmat
411000601
VILB411000601EA 279 EUR
VILB411000601
Négatoscopes, CN-6
Beleuchtung UV-Betrachtungsgeräte
The CN-6 darkroom holds one or two handheld UV lamps (VL-6 model) in any of the three wavelengths - 254 nm, 312 nm or 365 nm.
  • Black rubber curtain for easy access into the darkroom
  • UV absorber shield to protect the user from UV light
  • Removable lamps that can be used for handheld applications
W×D×H: 300×280×240 mm
Bestellinformation: To be completed with VL-6 lamps.
Lieferumfang: Supplied without lamps and allows different lighting possibilities according to the users choice.