Laboratory bottles, VITgrip™
Lieferant: VITLAB
110294 110194 110394 110494 110594
VITL110294EA 40.5 EUR
VITL110294 VITL110194 VITL110394 VITL110494 VITL110594
Laboratory bottles, VITgrip™
Flaschen Kunststoffflaschen
Made of PP, translucent, with tamper-evident closures.
  • With graduation
  • Excellent handling
  • Leakproof and break resistant
  • Safe storage
  • Robust and very versatile
  • Excellent chemical and temperature resistance
The laboratory bottle VITgrip™ is the allround bottle for storage and sample collection of liquids in the lab. The innovative design is characterised, amongst other things, by optimum grip and high accuracy of the graduation (±5%) and provides safe protection against leakage of liquids.
Standards: Suitable for contact with foodstuffs (according to regulation EU No. 10/2011).