Magnetic stirring bar retrievers, PTFE
Lieferant: VITLAB
122097
VITL122097EA 259 EUR
VITL122097
Magnetic stirring bar retrievers, PTFE
Rührer Rührwerkzeuge
With PTFE encapsulated magnetic core. Straight shape. High chemical resistance, simple to clean.