Sealing tape

Lieferant: VITLAB
131097
VITL131097EA 32.1 EUR
VITL131097
Sealing tape
Klebebänder
PTFE sealing tape for sealing and wrapping of threads and other connections.

  • High chemical resistance
  • Working temperatures up to + 250 °C

Length: 12,0 m

Colour: White
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR