Pipette controller, VITLAB pipeo®
Lieferant: VITLAB
1631500
VITL1631500EA 452 EUR
VITL1631500
Pipette controller, VITLAB pipeo®
Pipetten Pipettierhelfer
This pipette controller holds pipettes from 0,1 to 200 ml securely and tightly in the exchangeable adapter. Liquid vapours are purged directly to protect the instrument.
  • Ergonomic handle – very lightweight at about 190 grams – and excellent balance
  • Speed can be adjusted easily, continuously and exactly with one hand using two buttons
  • 50 ml pipette can be filled comfortably in less than ten seconds
  • Liquid release can be done either by gravity delivery when calibrated 'Ex' (to deliver), or in blow out mode using the battery-operated motor
  • One full charge of the nickel-metal hydride battery allows 8 hours of non-stop pipetting
  • Charge level of the recyclable battery is shown by the LED indicator
  • Pipette controller can be used while the battery is being recharged
Lieferumfang: Supplied with battery charger (100 to 240 V, 50/60 Hz), four plug adapters (EU, UK, US/J, AUS), battery, battery compartment cover, two replacement 0,2 µm membrane filters and instruction manual.