Magnetic stirring bar retriever, flexible, PTFE
Lieferant: VITLAB
318597
VITL318597EA 584 EUR
VITL318597
Magnetic stirring bar retriever, flexible, PTFE
Rührer Rührwerkzeuge
Flexible PTFE magnetic stirring bar retriever.
  • High chemical resistance
  • Magnet encapsulated (Ø×L: 12,5×51 mm)
  • Easy to clean