Tubing connectors
Lieferant: VITLAB
80535
VITL80535EA 0 EUR
VITL80535
Tubing connectors
Fittings und Verbinder
Connectors, made of HDPE, translucent, with tapered ends. Two connecting parts.
  • Good chemical resistance
  • Good thermal resistance (operating temperature up to 80 °C)
  • Unbreakable
Provides an excellent seal, but can be released easily at any time. Not suitable for partial vacuums. HDPE, useful for joining tubing of glass, rubber or plastic.