Wash bottles, narrow or wide neck
Lieferant: VITLAB
95188 94688 94188
VITL95188EA 99 EUR
VITL95188 VITL94688 VITL94188
Wash bottles, narrow or wide neck
Flaschen Spritzflaschen
Made of LDPE, translucent, screw cap and spray tube from PP.
  • Good chemical resistance
  • Good temperature resistance
Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.