Ultrapure water systems, arium® comfort I

Lieferant: Sartorius
H2O-I-2-TOC-B H2O-I-2-TOC-T H2O-I-2-UV-B H2O-I-1-B H2O-I-1-T H2O-I-1-TOC-B H2O-I-1-TOC-T H2O-I-1-UV-B H2O-I-1-UV-T H2O-I-2-B H2O-I-2-T H2O-I-2-UV-T
VIVAH2O-I-2-TOC-BEA 11440 EUR
VIVAH2O-I-2-TOC-B VIVAH2O-I-2-TOC-T VIVAH2O-I-2-UV-B VIVAH2O-I-1-B VIVAH2O-I-1-T VIVAH2O-I-1-TOC-B VIVAH2O-I-1-TOC-T VIVAH2O-I-1-UV-B VIVAH2O-I-1-UV-T VIVAH2O-I-2-B VIVAH2O-I-2-T VIVAH2O-I-2-UV-T
Ultrapure water systems, arium® comfort I
Wasseraufbereitungssysteme Reinstwassersysteme
It is compact, environmentally friendly, reliable and easy-to-use. arium® comfort I produces ASTM Type 1 ultrapure water and Type 3 pure water. The system contains state of the art reverse osmosis technology and a unique cartridge. arium® comfort I optimizes water consumption using the integrated iJust control unit. Applied for HPLC, GC-MS, AAS, ICP-MS, Ion chromatography, TOC-Analysis, photometry and for buffer solutions.

  • Optional integrated TOC monitor
  • Compact design
  • Flexible display and SD card slot
  • Feed water for laboratory devices such as autoclaves
More Product Information
Bestellen

TECHNISCHE DATEN

Modell arium® comfort I, with UV lamp and TOC monitor arium® comfort I, with UV lamp arium® comfort I, without UV lamp and TOC monitor arium® comfort I, with UV lamp and TOC monitor arium® comfort I, with UV lamp arium® comfort I, without UV lamp and TOC monitor
Environmentally Preferable
Fließrate 0,266 l/min 0,266 l/min 0,133 l/min 0,133 l/min 0,133 l/min 0,266 l/min
TOC-Gehalt ≤2 ppb ≤2 ppb ≤2 ppb ≤2 ppb ≤2 ppb ≤2 ppb
Spezifischer Widerstand bei +25 °C 18,2 MΩ.cm 18,2 MΩ.cm 18,2 MΩ.cm 18,2 MΩ.cm 18,2 MΩ.cm 18,2 MΩ.cm
Partikel <1/ml <1/ml <1/ml <1/ml <1/ml <1/ml
Mikroorganismen <1 CFU/1000 ml <1 CFU/1000 ml <1 CFU/1000 ml <1 CFU/1000 ml <1 CFU/1000 ml <1 CFU/1000 ml
B×T×H 435×476×501 mm 435×476×501 mm 435×476×501 mm 435×476×501 mm 435×476×501 mm 435×476×501 mm
Betriebsgewicht mit vollem Tank ~31 kg ~31 kg ~31 kg ~31 kg ~31 kg ~31 kg
Gewicht ~23 kg ~23 kg ~23 kg ~23 kg ~23 kg ~23 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR