Ultrapure water systems, arium® comfort II
Lieferant: Sartorius
H2O-II-2-T H2O-II-2-UV-T H2O-II-2-TOC-T H2O-II-2-TOC-B H2O-II-2-B H2O-II-2-UV-B H2O-II-1-TOC-T H2O-II-1-TOC-B H2O-II-1-T H2O-II-1-B H2O-II-1-UV-T H2O-II-1-UV-B
VIVAH2O-II-2-TEA 15330 EUR
VIVAH2O-II-2-T VIVAH2O-II-2-UV-T VIVAH2O-II-2-TOC-T VIVAH2O-II-2-TOC-B VIVAH2O-II-2-B VIVAH2O-II-2-UV-B VIVAH2O-II-1-TOC-T VIVAH2O-II-1-TOC-B VIVAH2O-II-1-T VIVAH2O-II-1-B VIVAH2O-II-1-UV-T VIVAH2O-II-1-UV-B
Ultrapure water systems, arium® comfort II
Wasseraufbereitungssysteme Reinstwassersysteme
The arium® comfort II system supplies ASTM Type I and Type II ultrapure water and is used for both critical applications and routine analyses. Streamlined to incorporate only the most important functions for the production of high quality ultrapure water, the arium® comfort II is a cost effective alternative for the discerning user that is also very easy to use.
  • Available in benchtop or wall mounted versions
  • Compact design, flexibility of use and simplicity of maintenance
  • Innovative glass operator interface with touch function
  • Time controlled dispensing
More Product Information