Wasser, Molekularbiologische Qualität nukleasen-, DNAse-, RNAse-frei, steril

Lieferant: VWR Chemicals

VQ65052.0100EA 26.9 EUR
VQ65052.0100 VQ65052.0500 VQ65052.1000
Wasser, Molekularbiologische Qualität nukleasen-, DNAse-, RNAse-frei, steril
Wasser

Molecular Biology grade water, nuclease-free is suitable for use in polymerase chain reaction and other molecular biology techniques requiring sterile nuclease-free conditions, such as in processing DNA or RNA (i.e., PCR, cDNA synthesis, qPCR or NGS Library).


  • Free of DNase activity (endonuclease, nicking and exonuclease activity)
  • Free of RNase and protease activity
  • Absence of human DNA, is tested by PCR
  • Highly purified, ultra-filtered and tested sterile
  • Not DEPC-treated water


Use this product as directed in the protocol of choice.


Bitte beachten Stable until expiry date as stated on the label.

Formel: H₂O
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Sterile (Bioburden test) PASS
DNAses free NONE
RNAses free NONE
Proteases free NONE
Human DNA free NONE


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR