Escherichia coli WDCM 00013 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Lieferant: Merck
Vitroids™ discs contain Escherichia coli WDCM 00013 viable microorganisms in a certified quantity (generally accredited according to ISO/IEC 17025), produced under reproducible conditions compliant with ISO 17034:2016 using authenticated strains from CECT®.
- Defined CFU range and low standard deviation
- Fast, reliable and easy to use
The discs consist of pure cultures of bacteria in a solid matrix, stable for at least one year and are in a viable state with a shelf life of 1-3 years.
Each batch is provided with a comprehensive certificate of analysis that specifies the mean number of colony-forming units (CFU), an expanded uncertainty about the mean, details about the method used to determine the product data and the number of passages (subcultures) from the original strain.
Vitroids™ are derived from a traceable culture obtained freeze-dried from CECT®.
Standards: These certified reference materials (CRMs) are produced and certified in accordance with ISO 17034 and ISO/IEC 17025.