Schwefelsäure 96% VLSI, elektronische Qualität
Lieferant: VWR Chemicals
|
Gefahr
|
VWRC6682.2500EA
169
EUR
VWRC6682.2500
Schwefelsäure 96% VLSI, elektronische Qualität
Schwefelsäure
Formel:
H₂SO₄ Molecular Weight: 98,08 g/mol Siedepunkt: 330 °C (1013 hPa) Schmelzpunkt: 10,38 °C Dichte: 1,84 kg/l Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064589 CAS-Nummer: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8,II REACH: 01-2119458838-20 Merck Index: 14,08974 |
Spezifikation Testergebnisse
Assay (acidimetric) | 96.0 - 97.0 % |
Colouration | ≤ 10 APHA |
Particle size(>0.50 μm)(/ml)(at filling) | ≤ 64 |
Cl (Chloride) | ≤ 100 ppb |
NH₄ (Ammonium) | ≤ 500 ppb |
NO₃ (Nitrate) | ≤ 200 ppb |
PO₄ (Phosphate) | ≤ 200 ppb |
Ag (Silver) | ≤ 10 ppb |
Al (Aluminium) | ≤ 20 ppb |
As + Sb (as As) | ≤ 5 ppb |
Au (Gold) | ≤ 20 ppb |
B (Boron) | ≤ 10 ppb |
Ba (Barium) | ≤ 10 ppb |
Be (Beryllium) | ≤ 10 ppb |
Bi (Bismuth) | ≤ 20 ppb |
Ca (Calcium) | ≤ 100 ppb |
Cd (Cadmium) | ≤ 10 ppb |
Co (Cobalt) | ≤ 10 ppb |
Cr (Chromium) | ≤ 10 ppb |
Cu (Copper) | ≤ 10 ppb |
Fe (Iron) | ≤ 50 ppb |
Ga (Gallium) | ≤ 10 ppb |
Ge (Germanium) | ≤ 10 ppb |
In (Indium) | ≤ 10 ppb |
K (Potassium) | ≤ 50 ppb |
Li (Lithium) | ≤ 10 ppb |
Mg (Magnesium) | ≤ 20 ppb |
Mn (Manganese) | ≤ 10 ppb |
Mo (Molybdenum) | ≤ 10 ppb |
Na (Sodium) | ≤ 50 ppb |
Ni (Nickel) | ≤ 10 ppb |
Pb (Lead) | ≤ 10 ppb |
Pt (Platinum) | ≤ 20 ppb |
Se (Selenium) | ≤ 100 ppb |
Sn (Tin) | ≤ 20 ppb |
Sr (Strontium) | ≤ 10 ppb |
Ti (Titanium) | ≤ 10 ppb |
Tl (Thallium) | ≤ 20 ppb |
V (Vanadium) | ≤ 10 ppb |
Zn (Zinc) | ≤ 50 ppb |
Zr (Zirconium) | ≤ 10 ppb |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...