HMDS (1,1,1,3,3,3-Hexamethyldisilazan) ≥99,5% VLSI, elektronische Qualität

Lieferant: VWR Chemicals
Gefahr

VWRC6686.2500EA 524 EUR
VWRC6686.2500
HMDS (1,1,1,3,3,3-Hexamethyldisilazan) ≥99,5% VLSI, elektronische Qualität
HMDS (1,1,1,3,3,3-Hexamethyldisilazan)
Formel: (CH₃)₃SiNHSi(CH₃)₃
Dichte: 0,78 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008259
CAS-Nummer: 999-97-3
EINECS: 213-668-5
UN: 1992
ADR: 3 (6.1, 8),II

Bestellen

Spezifikation Testergebnisse

Assay (GC) ≥ 99.5 %
Impurity profile (GC) Conforms
Colouration ≤ 10 APHA
Particle size(0.50 μm)(/ml)(at filling) ≤ 200
Evaporation residue ≤ 10 ppm
Water ≤ 500 ppm
Cl (Chloride) ≤ 1000 ppb
Ag (Silver) ≤ 10 ppb
Al (Aluminium) ≤ 100 ppb
As (Arsenic) ≤ 10 ppb
Au (Gold) ≤ 20 ppb
B (Boron) ≤ 10 ppb
Ba (Barium) ≤ 20 ppb
Be (Beryllium) ≤ 10 ppb
Bi (Bismuth) ≤ 20 ppb
Ca (Calcium) ≤ 100 ppb
Cd (Cadmium) ≤ 10 ppb
Co (Cobalt) ≤ 10 ppb
Cr (Chromium) ≤ 10 ppb
Cu (Copper) ≤ 10 ppb
Fe (Iron) ≤ 50 ppb
Ga (Gallium) ≤ 10 ppb
In (Indium) ≤ 10 ppb
K (Potassium) ≤ 20 ppb
Li (Lithium) ≤ 10 ppb
Mg (Magnesium) ≤ 20 ppb
Mn (Manganese) ≤ 10 ppb
Mo (Molybdenum) ≤ 10 ppb
Na (Sodium) ≤ 100 ppb
Ni (Nickel) ≤ 10 ppb
Pb (Lead) ≤ 10 ppb
Pt (Platinum) ≤ 50 ppb
Sb (Antimony) ≤ 10 ppb
Sn (Tin) ≤ 20 ppb
Sr (Strontium) ≤ 10 ppb
Ti (Titanium) ≤ 20 ppb
Tl (Thallium) ≤ 10 ppb
V (Vanadium) ≤ 10 ppb
Zn (Zinc) ≤ 20 ppb
Zr (Zirconium) ≤ 20 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR