E/E 70% v/v, KLINIPATH
Lieferant: KLINIPATH
Synonyme:
Ethyl alcohol
4070-9001
4070-9010
VWRK4070-9001EA
65.3
EUR
VWRK4070-9001
VWRK4070-9010
E/E 70% v/v, KLINIPATH
Ethanol
Formel:
C₂H₅OH Molecular Weight: 46.07 g/mol Siedepunkt: 78.3 Schmelzpunkt: -117 Flammpunkt: 12 |
MDL:
MFCD00003568 CAS-Nummer: 64-17-5 UN: 1170 ADR: 3 III Merck Index: 12,03806 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...