Wasser, AnalaR NORMAPUR® ISO 3696 Grade 3 zA
Lieferant: VWR Collection
Synonyme:
Wasserstoffoxid, Hydrogenoxid
VWRS10292MBEA
0
EUR
VWRS10292MB
Wasser, AnalaR NORMAPUR® ISO 3696 Grade 3 zA
Wasser
Produziert durch Reversosmose und CDI
Formel:
H₂O Molecular Weight: 18.02 g/mol Siedepunkt: 100 °C (1013 hPa) Schmelzpunkt: 0 °C Dichte: 1 g/cm³ (4 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Spezifikation Testergebnisse
Substances reducing KMnO4 | Passes test |
Conductivity (at filling) | Max. 1 µS/cm |
pH (25°C) | 5.0 - 7.5 |
Evaporation residue | Max. 1 ppm |
Ignition residue | Max. 1 ppm |
Silica | Max. 0.1 ppm |
Cl (Chloride) | Max. 0.05 ppm |
F (Fluoride) | Max. 0.05 ppm |
NH4 (Ammonium) | Max. 0.2 ppm |
NO3 (Nitrate) | Max. 0.01 ppm |
PO4 (Phosphate) | Max. 0.01 ppm |
SO4 (Sulphate) | Max. 0.1 ppm |
Ag (Silver) | Max. 0.0004 ppm |
Al (Aluminium) | Max. 0.002 ppm |
As (Arsenic) | Max. 0.002 ppm |
Au (Gold) | Max. 0.001 ppm |
B (Boron) | Max. 0.005 ppm |
Ba (Barium) | Max. 0.001 ppm |
Be (Beryllium) | Max. 0.002 ppm |
Bi (Bismuth) | Max. 0.001 ppm |
Ca (Calcium) | Max. 0.05 ppm |
Cd (Cadmium) | Max. 0.001 ppm |
Co (Cobalt) | Max. 0.001 ppm |
Cr (Chromium) | Max. 0.0004 ppm |
Cu (Copper) | Max. 0.0004 ppm |
Fe (Iron) | Max. 0.001 ppm |
Ga (Gallium) | Max. 0.004 ppm |
Ge (Germanium) | Max. 0.001 ppm |
In (Indium) | Max. 0.001 ppm |
K (Potassium) | Max. 0.05 ppm |
Li (Lithium) | Max. 0.002 ppm |
Mg (Magnesium) | Max. 0.005 ppm |
Mn (Manganese) | Max. 0.0004 ppm |
Mo (Molybdenum) | Max. 0.002 ppm |
Na (Sodium) | Max. 0.1 ppm |
Ni (Nickel) | Max. 0.0004 ppm |
Pb (Lead) | Max. 0.001 ppm |
Pd (Palladium) | Max. 0.008 ppm |
Pt (Platinum) | Max. 0.001 ppm |
Sb (Antimony) | Max. 0.001 ppm |
Sn (Tin) | Max. 0.001 ppm |
Sr (Strontium) | Max. 0.0004 ppm |
Ti (Titanium) | Max. 0.001 ppm |
Tl (Thallium) | Max. 0.00005 ppm |
V (Vanadium) | Max. 0.001 ppm |
Zn (Zinc) | Max. 0.004 ppm |
Zr (Zirconium) | Max. 0.001 ppm |
ISO Standard Water Grade 3 | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...