Gummi Xanthan
Lieferant: VWR Collection
Synonyme:
Xanthan Gummi, Xanthan
VWRS5859.5000EA
0
EUR
VWRS5859.5000
Gummi Xanthan
Xanthan gum
Dichte:
1,5 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00131256 CAS-Nummer: 11138-66-2 EINECS: 234-394-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...