4-(Dimethylamino)pyridin 14,5 g/l in THF, VOLUSOL®

Lieferant: VWR Collection
Gefahr

VWRS83927.410EA 0 EUR
VWRS83927.410
4-(Dimethylamino)pyridin 14,5 g/l in THF, VOLUSOL®
4-(Dimethylamino)pyridin
Formel: C₇H₁₀N₂
MDL: MFCD00006418
CAS-Nummer: 1122-58-3
UN: 2056
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR