BioCleanTip Wako® Micropipette Tips
Lieferant: FUJIFILM WAKO CHEMICALS
298-35031 291-35021 294-35011
WAKF298-35031EA 75.4 EUR
WAKF298-35031 WAKF291-35021 WAKF294-35011
BioCleanTip Wako® Micropipette Tips
Pipettenspitzen
BioClean BioCleanTip Wako® are endotoxin-free micropipette tips that are individually wrapped to reduce the chance of contamination and are perfect for all types of LAL testing.
  • Endotoxin-free pipette tip (<0,005 EU/tip)
  • Individually packaged, ideal for use in clean rooms
  • Packaging film features dust-free protection
  • Pipette tips fit most micropipettes
The BioCleanTip Wako® are available in both 200 µl and 1000 µl volumes, with the 200 µl volume tips being offered in standard and extended sizes. The BioCleanTip Wako® Extend S are to can easilyMore Product Information