Accessories for Endotoxin Tests
Lieferant: FUJIFILM WAKO CHEMICALS
293-51601
WAKF293-51601EA 309 EUR
WAKF293-51601
Accessories for Endotoxin Tests
Mikrobiologische Testsysteme Endotoxin-Tests
This endotoxin-free solution containing human serum albumin (HSA) is capable of extracting endotoxins that cannot be extracted in water or saline solution.
Traditionally, water or saline solution has been used to extract endotoxin in tests involving medical devices and equipment; however, the efficacy of this extraction method is debated. By using this extraction solution, it provides our customers with a more reliable method for medical device testing.
Recommended for use in endotoxin testing on equipment and devices that may come into contact with fluids containing blood or protein.