αBET™ CMD Rapid Endotoxin Detection System
Lieferant: FUJIFILM WAKO CHEMICALS
CMD-A-0006-C CMD-A-0007-EN
WAKFCMD-A-0006-CEA 691 EUR
WAKFCMD-A-0006-C WAKFCMD-A-0007-EN
αBET™ CMD Rapid Endotoxin Detection System
Mikrobiologische Testsysteme Endotoxin-Tests
The CMD αBET™ system is a fully integrated testing system that provides a rapid and sustainable solution to endotoxin testing without compromising on sensitivity or performance. Using our FDA-licensed turbidimetric Limulus amebocyte lysate (LAL) reagent, quantitative results are provided in under half the time of other testing formats, with less reagent consumption and minimal user input.
  • Data back up to PC by binary code, preventing the ability of data modification
  • Backups made in two formats-native firmware and HTML for ease of viewing records
  • 7" touchscreen
  • QR code access cards included (1 administrator card, 10 user cards)
  • Run times of no more than 30 minutes per assay can achieve 0,001 EU/ml
Uses less reagent than traditional methods. Minimal user input needed beyond addition of sample. Guided steps for performing various assays. Compatible with color and opaque samples.
Portable and fully integrated bench top system with FDA 21 CFR Part 11 compliant software.
For use with PYROSTAR™ ES-F 0,015 EU/ml reagent available in single test, 20 test, and 50 test vials.