Depyrogenated Dilution Tubes
Lieferant: FUJIFILM WAKO CHEMICALS
These depyrogenated dilution tubes are made of high-quality borosilicate glass depyrogenated at 250 °C.
- Endotoxin-free test tubes (<0,001 EU/tube)
- Designed for all testing methods
- Made with borosilicate glass
- Can be used for general lab testing
This results in endotoxin-free tubes that can perform successful LAL dilution series, from creating the standard curve to sample dilutions, with minimal chance of potential contamination.