Accessories for Endotoxin Tests
Lieferant: FUJIFILM WAKO CHEMICALS
LRW-12125
LRW-2030
WAKFLRW-12125EA
439
EUR
WAKFLRW-12125
WAKFLRW-2030
Accessories for Endotoxin Tests
Mikrobiologische Testsysteme
Endotoxin-Tests
LAL Reagent Water can be used to fulfill all aspects of endotoxin testing. From reconstitution of both the CSE and LAL for the creation of standard curves to the making of product dilutions, LAL reagent water provides reliable and reproducible data when paired with our endotoxin-specific LAL reagents.
- Endotoxin-free water (<0,001 EU/ml)
- Great for reconstitution of LAL, RSE and CSE
- Used to perform dilution series of RSE and CSE
- Used for sample dilutions