LPS for Endotoxin Indicator
Lieferant: FUJIFILM WAKO CHEMICALS
WLPS-0100K
WAKFWLPS-0100KEA
353
EUR
WAKFWLPS-0100K
LPS for Endotoxin Indicator
Mikrobiologische Testsysteme
Endotoxin-Tests
The LPS for Endotoxin Indicator is composed of E. coli O55:B5 (>100,000 EU/vial) and serves as a stock solution to prepare multiple endotoxin indicators for depyrogenation validation studies.
- E. coli 055:B5 (>100,000 EU/vial)
- Can make multiple endotoxin indicators from one vial
- Used for dry heat oven validation studies
The prepared endotoxin indicators may be tested using reagents from our PYROSTAR™ ES-F series.