Digitonin

Lieferant: Wako

Synonyme: Digitonin

047-21374
WAKO047-21374EA 10360 EUR
WAKO047-21374
Digitonin
Digitonin
Formel: C₅₆H₉₂O₂₉
Molecular Weight: 1229.312 g/mol
MDL: MFCD00077729
CAS-Nummer: 11024-24-1
EINECS: 234-255-6
UN: 3462
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR