Molekularsieb 13X (1,0 nm, 10 Å)

Lieferant: Wako

131-07085
WAKO131-07085EA 0 EUR
WAKO131-07085
Molekularsieb 13X (1,0 nm, 10 Å)
Molekularsieb 13X (1,0 nm, 10 Å)
MDL: MFCD00164506
CAS-Nummer: 63231-69-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR