Xylencyanol FF
Lieferant: Wako
|
Achtung
|
Synonyme:
Acid Blue 147, Hydrogen-4-[[4-(ethylamino)-m-tolyl][4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-yliden]methyl]benzol-1,3-disulfonatnatrium
242-00462
WAKO242-00462EA
446
EUR
WAKO242-00462
Xylencyanol FF
Xylencyanol FF
Formel:
C₂₅H₂₇N₂NaO₆S₂ Molecular Weight: 538,62 g/mol Schmelzpunkt: 295 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040657 CAS-Nummer: 2650-17-1 EINECS: 220-167-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...