2-Butyl-1,3-propandiol

Lieferant: Wako

Synonyme: 2-Butylpropane-1,3-diol

323-31051
WAKO323-31051EA 168 EUR
WAKO323-31051
2-Butyl-1,3-propandiol
2-Butyl-1,3-propanediol
Formel: C₇H₁₆O₂
Molecular Weight: 132,2 g/mol
MDL: MFCD00799395
CAS-Nummer: 2612-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR