2-Methylthiazol

Lieferant: Wako

326-44552-25G
WAKO326-44552-25GEA 283 EUR
WAKO326-44552-25G
2-Methylthiazol
2-Methylthiazole
Formel: C₄H₅NS
Molecular Weight: 99,16 g/mol
MDL: MFCD00053144
CAS-Nummer: 3581-87-1
EINECS: 222-702-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR